Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056626 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056626 0.2 mL
NB10056626T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056626 Supplier Novus Biologicals Supplier No. NB10056626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIM1 Polyclonal specifically detects LIM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen LIM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_005559
Gene Alias hLim-1, Homeobox protein Lim-1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723
Gene Symbols LHX1
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbit with synthetic peptides corresponding to amno acids 149-165, 175-162, and 395-412 of human Lim-1.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3975
Test Specificity In human muscle, a 60 kDa band is observed.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.