Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM1 Antibody (OTI2D5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP271134C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271134C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271134C Supplier Novus Biologicals Supplier No. NBP271134C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIM1 Monoclonal specifically detects LIM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LIM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2D5
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias hLim-1, Homeobox protein Lim-1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723
Gene Symbols LHX1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 100-362 of human LIM1 (NP_005559) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3975.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.