Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB159609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159609 25 μg
NB159608 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159609 Supplier Novus Biologicals Supplier No. NBP31778925UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIM1 Polyclonal antibody specifically detects LIM1 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen LIM1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias hLim-1, Homeobox protein Lim-1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: FVCKEDYLSNSSVAKENSLHSATTGSDPSLSPDSQDPSQDDAKDSESANVSDKEAGSNENDDQNL
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3975
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.