Learn More
Description
Specifications
Specifications
| Antigen | LIM2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | lens fiber membrane intrinsic protein, lens intrinsic membrane protein 2, 19kDa, MP17, MP18, MP19lens intrinsic membrane protein 2 (19kD), MP20 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 25-110 of human LIM2 (NP_085915.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
