Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME Antibody (LIME-06) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP144945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144945 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144945 Supplier Novus Biologicals Supplier No. NBP144945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIME Monoclonal specifically detects LIME in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen LIME
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone LIME-06
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunoprecipitation 1:50
Gene Alias dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Gene Symbols LIME1
Host Species Mouse
Immunogen Bacterially expressed intracellular fragment corresponding to aa 141-295 of human LIME.
Molecular Weight of Antigen 30 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54923
Test Specificity The antibody LIME-06 was raised against intracellular fragment corresponding to aa 141-295 of human LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed by T cells.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.