Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME Antibody (mLIME-05), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP147230AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147230AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147230AH Supplier Novus Biologicals Supplier No. NBP147230AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIME Monoclonal specifically detects LIME in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LIME
Applications Western Blot
Classification Monoclonal
Clone mLIME-05
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Gene Symbols LIME1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 86-202 of mouse LIME
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54923
Test Specificity Mouse monoclonal antibody mLIME-05 reacts with the cytoplasmic domain of LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed mainly by T cells.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.