Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME Antibody (OTI5D3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271974UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271974UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271974UV Supplier Novus Biologicals Supplier No. NBP271974UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIME Monoclonal specifically detects LIME in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LIME
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5D3
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Gene Symbols LIME1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 30-295 of human LIME (NP_060276) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54923.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.