Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Limestone Specimen
SCI_ED

Catalog No. S26591
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Limestone Specimen is made up of mostly calcium carbonate and organic material.

  • Limestone formed from precipitation of calcium carbonate out of solution, in ocean or in shallow marine environment
  • For use in demonstrating tendency of calcium carbonate to effervesce in acid
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Massive, gray variety is often found deposited in marine environments
  • Limestone is important rock in industry and education

Specifications

Product Type Sedimentary Rock, Limestone
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.