Learn More
Eisco™ Limonite Mineral Specimen
SCI_ED
Catalog No. p-7203006
Click to view available options
Quantity:
1/Ea.
12/Pk.
Limonite, also known as goethite, is an iron hydroxide mineral, a chemistry that is apparent from the mineral's rusty ochre color. This mineral was used extensively as pigment for yellow, red, and orange dyes, going back all the way to Neolithic cave paintings.
  • Correlates well to Next Generation Science Standards Evidence
  • Useful for lessons in mineral chemistry and identification
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.