Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMPII/SR-B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 12 publications
Supplier: Novus Biologicals NB400102
Description
LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
LIMPII/SR-B2 | |
Polyclonal | |
Western Blot 1:1000, Flow Cytometry reported in scientific literature, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:25-1:100, Immunohistochemistry-Paraffin 1:25, KnockDown Validated reported in scientific literature (PMID 31132962) | |
O35114 | |
SCARB2 | |
A peptide containing residues from mouse SR-BII (between residues 400-478) plus an N-terminal cysteine was coupled to KLH. [UniProt# O35114] | |
Unpurified | |
RUO | |
Primary | |
Mouse, rat, bovine, human, mink and Chinese hamster. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.1% Sodium Azide | |
AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII | |
Rabbit | |
82 kDa | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
950 | |
Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction