Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIMPII/SR-B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 12 publications
$206.50 - $501.50
Specifications
Antigen | LIMPII/SR-B2 |
---|---|
Dilution | Western Blot 1:1000, Flow Cytometry reported in scientific literature, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:25-1:100, Immunohistochemistry-Paraffin 1:25, KnockDown Validated reported in scientific literature (PMID 31132962) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400102SS
![]() |
Novus Biologicals
NB400102SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400102
![]() |
Novus Biologicals
NB400102 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
LIMPII/SR-B2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
Whole antisera with 0.1% Sodium Azide | |
AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII | |
SCARB2 | |
IgG | |
Unpurified | |
Mouse, rat, bovine, human, mink and Chinese hamster. |
Western Blot 1:1000, Flow Cytometry reported in scientific literature, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:25-1:100, Immunohistochemistry-Paraffin 1:25, KnockDown Validated reported in scientific literature (PMID 31132962) | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid | |
O35114 | |
950 | |
A peptide containing residues from mouse SR-BII (between residues 400-478) plus an N-terminal cysteine was coupled to KLH. [UniProt# O35114] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
82 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title