Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMPII/SR-B2 Antibody, Novus Biologicals™
SDP

Catalog No. NB400102SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400102SS 0.025 mL
NB400102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400102SS Supplier Novus Biologicals Supplier No. NB400102SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 12 publications

LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen LIMPII/SR-B2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry reported in scientific literature, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:25-1:100, Immunohistochemistry-Paraffin 1:25, KnockDown Validated reported in scientific literature (PMID 31132962)
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. O35114
Gene Alias AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII
Gene Symbols SCARB2
Host Species Rabbit
Immunogen A peptide containing residues from mouse SR-BII (between residues 400-478) plus an N-terminal cysteine was coupled to KLH. [UniProt# O35114]
Molecular Weight of Antigen 82 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 950
Test Specificity Mouse, rat, bovine, human, mink and Chinese hamster.
Target Species Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.