Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMPII/SR-B2 Antibody, Novus Biologicals™
SDP

Catalog No. NB400129SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400129SS 0.025 mL
NB400129 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400129SS Supplier Novus Biologicals Supplier No. NB400129SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 17 publications

LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen LIMPII/SR-B2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23863627)
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII
Gene Symbols SCARB2
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to the mouse LIMPII/lgp85 protein sequence. [UniProt# O35114]
Molecular Weight of Antigen 85 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 950
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.