Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN-28A Rabbit anti-Human, Mouse, Rat, Clone: JA10-63, Novus Biologicals™
SDP

Catalog No. NBP266932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266932 Supplier Novus Biologicals Supplier No. NBP266932

Rabbit Monoclonal Antibody

LIN-28A Monoclonal antibody specifically detects LIN-28A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin).

Specifications

Antigen LIN-28A
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JA10-63
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunoprecipitation 1:10-1:50, Immunohistochemistry-Paraffin 1:400
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1
Gene Symbols LIN28A
Host Species Rabbit
Immunogen Synthetic peptide within Human LIN-28A aa 10-59/ 209. (SwissProt: Q9H9Z2 Human; SwissProt: Q8K3Y3 Mouse; SwissProt: D3ZZA6 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 79727
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.