Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN-28B Mouse anti-Human, Clone: A2-4-5, Novus Biologicals™
SDP

Catalog No. NBP266933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266933 Supplier Novus Biologicals Supplier No. NBP266933

Mouse Monoclonal Antibody

LIN-28B Monoclonal specifically detects LIN-28B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LIN-28B
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A2-4-5
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CSDD2protein lin-28 homolog B, FLJ16517, lin-28 homolog B (C. elegans), Lin-28.2, lin-28B
Gene Symbols LIN28B
Host Species Mouse
Immunogen Recombinant protein within Human LIN-28B aa 73-250/ 250. (SwissProt: Q6ZN17 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 389421
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.