Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN52 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB351185 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351185 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351185 Supplier Novus Biologicals Supplier No. NBP306592MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIN52 Polyclonal antibody specifically detects LIN52 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen LIN52
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias c14_5549, C14orf46, chromosome 14 open reading frame 46, lin-52 homolog (C. elegans), protein lin-52 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human LIN52 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91750
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.