Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINE-1 ORF1p Antibody, Novus Biologicals™
SDP

Catalog No. NBP266934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266934 Supplier Novus Biologicals Supplier No. NBP266934

Rabbit Polyclonal Antibody

LINE-1 ORF1p Polyclonal specifically detects LINE-1 ORF1p in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LINE-1ORF1p
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias L LINE-1 retrotransposable element ORF1 protein, L1-ORF1p, LINE retrotransposable element 1, LINE1 retrotransposable element 1, Transposase element L1Md-A101/L1Md-A102/L1Md-A2
Host Species Rabbit
Immunogen Synthetic peptide within mouse LINE-1 ORF1p aa 308-357/ 357. (SwissProt: P11260 Mouse)
Purification Method Immunogen affinity purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.