Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO-1 Antibody (332237), FITC, Novus Biologicals™
SDP

Catalog No. NB363324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363324 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363324 Supplier Novus Biologicals Supplier No. FAB30861F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LINGO-1 Monoclonal specifically detects LINGO-1 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen LINGO-1
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 332237
Conjugate FITC
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias FLJ14594, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LINGO-1, Thr40-Thr556, Accession # NP_116197
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84894
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.