Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO-2 Antibody (382007), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312809 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312809 Supplier Novus Biologicals Supplier No. FAB36791PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LINGO-2 Monoclonal specifically detects LINGO-2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LINGO-2
Applications Flow Cytometry
Classification Monoclonal
Clone 382007
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LRRN6Cleucine rich repeat neuronal 6C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LINGO-2, Cys28-Leu542, Accession # Q7L985
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158038
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.