Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LINGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 190971AP150UL
Description
Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors. Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination.Specifications
| LINGO1 | |
| Polyclonal | |
| Unconjugated | |
| LINGO1 | |
| LERN1, LINGO1, LRRN6A, UNQ201 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 235402, 315691, 84894 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96FE5, Q9D1T0 | |
| Lingo1 | |
| LINGO1 Fusion Protein Ag13553 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction