Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LINGO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LINGO1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2339
![]() ![]() |
Proteintech Group Inc
190971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors. Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination.Specifications
LINGO1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q96FE5, Q9D1T0 | |
235402, 315691, 84894 | |
LINGO1 Fusion Protein Ag13553 | |
Primary | |
-20°C | |
LINGO1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LERN1, LINGO1, LRRN6A, UNQ201 | |
Lingo1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title