Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipase A Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB338208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338208 Supplier Novus Biologicals Supplier No. NBP154155PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lipase A Polyclonal antibody specifically detects Lipase A in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Lipase A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Acid cholesteryl ester hydrolase, Cholesteryl esterase, EC 3.1.1, EC 3.1.1.13, LALCESDcholesterol ester hydrolase, Lipase A, lipase A, lysosomal acid, cholesterol esterase, lysosomal acid lipase, lysosomal acid lipase/cholesteryl ester hydrolase, Sterol esterase
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal region of the human Lipase A protein (within residues 150-300). [Swiss-Prot P38571]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3988
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.