Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP21419425UL
Description
LIPH Polyclonal specifically detects LIPH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LIPH | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
ARWH2, EC 3.1.1, EC 3.1.1.-, EC 3.1.1.3, LAH2, lipase H, lipase member H, lipase, member H, LPD lipase-related protein, LPDLR, Membrane-associated phosphatidic acid-selective phospholipase A1-alpha, membrane-bound phosphatidic acid-selective phospholipase A1, mPA-PLA1, MPAPLA1, mPA-PLA1 alpha, Phospholipase A1 member B, PLA1BAH | |
Rabbit | |
Affinity Purified | |
RUO | |
200879 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
LIPH | |
This antibody was developed against a recombinant protein corresponding to the amino acids: GCPKTILGGFQYFKCDHQRSVYLYLSSLRESCTITAYPCDSYQDYRNGKCVSCGTSQKESCPLLGYYADNWKDHL | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction