Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094173 100 μL
NB094174 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094173 Supplier Novus Biologicals Supplier No. NBP303946100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIPH Polyclonal specifically detects LIPH in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen LIPH
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ARWH2, EC 3.1.1, EC 3.1.1.-, EC 3.1.1.3, LAH2, lipase H, lipase member H, lipase, member H, LPD lipase-related protein, LPDLR, Membrane-associated phosphatidic acid-selective phospholipase A1-alpha, membrane-bound phosphatidic acid-selective phospholipase A1, mPA-PLA1, MPAPLA1, mPA-PLA1 alpha, Phospholipase A1 member B, PLA1BAH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human LIPH (NP_640341.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200879
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.