Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | LIPH |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LIPH Polyclonal specifically detects LIPH in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LIPH | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
200879 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
ARWH2, EC 3.1.1, EC 3.1.1.-, EC 3.1.1.3, LAH2, lipase H, lipase member H, lipase, member H, LPD lipase-related protein, LPDLR, Membrane-associated phosphatidic acid-selective phospholipase A1-alpha, membrane-bound phosphatidic acid-selective phospholipase A1, mPA-PLA1, MPAPLA1, mPA-PLA1 alpha, Phospholipase A1 member B, PLA1BAH | |
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human LIPH (NP_640341.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title