Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipid A Antibody (43) - Free, Novus Biologicals™
SDP

Catalog No. NB119840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119840 Supplier Novus Biologicals Supplier No. NBP311215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lipid A Monoclonal specifically detects Lipid A in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Lipid A
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 43
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation Culture Medium
Host Species Mouse
Immunogen The monoclonal antibody 43 reacts with the hydrophilic backbone of free lipid A.
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.