Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipid A LPS Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017587 0.1 mL
NB10064484 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017587 Supplier Novus Biologicals Supplier No. NB100644840.1ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Lipid A LPS Polyclonal specifically detects Lipid A LPS in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lipid A LPS
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation Phosphate buffered saline with 0.1% Sodium Azide
Host Species Goat
Immunogen Whole cell preparation of lipid A from E. coli O157.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64484 recognizes the Lipid A or endotoxin region of LPS (Lipopolysaccharide) from E. coli. Lipid A is a glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria. This antibody cross reacts with P. aeruginosa, K. pneumoniae, S. enteriditis, E. aerogenes, E. hermanii, Y. enterocolitica and S. sonnei.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.