Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipid A LPS Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$192.94 - $519.67
Specifications
| Antigen | Lipid A LPS |
|---|---|
| Concentration | 4 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017587
![]() |
Novus Biologicals
NB100644840.1ML |
0.1 mL |
Each for $192.94
|
|
|||||
NB10064484
![]() |
Novus Biologicals
NB10064484 |
0.5 mL |
Each for $519.67
|
|
|||||
Description
Lipid A LPS Polyclonal specifically detects Lipid A LPS in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Lipid A LPS | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| IgG | |
| IgG purified | |
| NB100-64484 recognizes the Lipid A or endotoxin region of LPS (Lipopolysaccharide) from E. coli. Lipid A is a glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria. This antibody cross reacts with P. aeruginosa, K. pneumoniae, S. enteriditis, E. aerogenes, E. hermanii, Y. enterocolitica and S. sonnei. |
| 4 mg/mL | |
| Unconjugated | |
| Goat | |
| Bacteria | |
| Whole cell preparation of lipid A from E. coli O157. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title