Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipin 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339258 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339258 Supplier Novus Biologicals Supplier No. NB11057151MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lipin 1 Polyclonal antibody specifically detects Lipin 1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Lipin 1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp781P1796, EC 3.1.3.18, EC 3.1.3.4, EC 5.4.2.7, KIAA0188phosphatidate phosphatase LPIN1, lipin 1, lipin-1, PAP1
Host Species Rabbit
Immunogen A synthetic peptide to an internal region (within residues 500-600) of the human LPIN1 protein. [Swiss-Prot# Q14693]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23175
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.