Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipin 1 Antibody (OTI8F10), PE, Novus Biologicals™
SDP

Catalog No. NB288686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288686 Supplier Novus Biologicals Supplier No. NBP271726PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lipin 1 Monoclonal antibody specifically detects Lipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Lipin 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8F10
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DKFZp781P1796, EC 3.1.3.18, EC 3.1.3.4, EC 5.4.2.7, KIAA0188phosphatidate phosphatase LPIN1, lipin 1, lipin-1, PAP1
Host Species Mouse
Immunogen Full length human recombinant protein of human LPIN1 (NP_663731) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23175
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.