Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipin 3 Antibody (OTI1D7), Novus Biologicals™
SDP

Catalog No. NBP245672 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245672 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245672 Supplier Novus Biologicals Supplier No. NBP245672
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

Lipin 3 Monoclonal specifically detects Lipin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Lipin 3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2
Gene Symbols LPIN3
Host Species Mouse
Immunogen Full length human recombinant protein of human LPIN3 (NP_075047) produced in HEK293T cell.
Molecular Weight of Antigen 93.4 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64900
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.