Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Antibody (56), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343309 Supplier Novus Biologicals Supplier No. NBP241366MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Human, Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA

Specifications

Antigen Lipocalin-2/NGAL
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 56
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Host Species Mouse
Immunogen Recombinant monkey NGAL
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3934
Target Species Human, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.