Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Antibody (68), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP241368A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP241368A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP241368A32 Supplier Novus Biologicals Supplier No. NBP241368AF532

Mouse Monoclonal Antibody

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human, Monkey samples. It is validated for ELISA.

Specifications

Antigen Lipocalin-2/NGAL
Applications ELISA
Classification Monoclonal
Clone 68
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Gene Symbols LCN2
Host Species Mouse
Immunogen Recombinant monkey NGAL
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3934.0
Test Specificity Antibody reacts with NGAL (neutrophil gelatinase-associated lipocalin) from Macaca mulatta (Rhesus) and Macaca facicularis (Cynomolgus).
Target Species Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.