Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Antibody, Novus Biologicals™
SDP

Catalog No. NBP145682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145682 50 μg
NB018574 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145682 Supplier Novus Biologicals Supplier No. NBP145682

Rabbit Polyclonal Antibody

Lipocalin-2/NGAL Polyclonal specifically detects Lipocalin-2/NGAL in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lipocalin-2/NGAL
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 μg/mL, Simple Western, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.2), 30% glycerol, 0.5% BSA with 0.01% Thimerosal
Gene Accession No. P11672
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Gene Symbols LCN2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding amino acid 40 of mouse Lipocalin 2.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3934
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.