Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipolysis Stimulated Lipoprotein Receptor Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$428.79 - $684.12
Specifications
| Antigen | Lipolysis Stimulated Lipoprotein Receptor |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Lipolysis Stimulated Lipoprotein Receptor Polyclonal antibody specifically detects Lipolysis Stimulated Lipoprotein Receptor in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Lipolysis Stimulated Lipoprotein Receptor | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS, pH 7.2, 40% glycerol | |
| 51599 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ILDR3, immunoglobulin-like domain containing receptor 3, lipolysis stimulated lipoprotein receptor, lipolysis-stimulated lipoprotein receptor, lipolysis-stimulated receptor, lipolysis-stimulated remnant, LISCH, LISCH protein, LISCH7, liver-specific bHLH-Zip transcription factor, MGC10659, MGC48312, MGC48503 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GVPSIYAPSTYAHLSPAKTPPPPAMIPMGPAYNGYPGGYPGDVDRSSSAGGQGSYVPLLRDTDSSVASEVRSGYRIQASQQDDSM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title