Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipoprotein Lipase/LPL Antibody (OTI3A10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP271178A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271178A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271178A30 Supplier Novus Biologicals Supplier No. NBP271178AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lipoprotein Lipase/LPL Monoclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Lipoprotein Lipase/LPL
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3A10
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase
Gene Symbols LPL
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 28-475 of human Lipoprotein Lipase/LPL (NP_000228) produced in HEK293T.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4023.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.