Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lipoyltransferase 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen lipoyltransferase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB5491220UL
SDP
View Documents
Novus Biologicals
NBP15491220UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP154912
SDP
View Documents
Novus Biologicals
NBP154912
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

lipoyltransferase 1 Polyclonal specifically detects lipoyltransferase 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

lipoyltransferase 1
Polyclonal
Rabbit
Lipid and Metabolism
EC 2.3.1.-, Lipoyl ligase, lipoyltransferase 1, mitochondrial
LIPT1
IgG
42 kDa
Western Blot
Unconjugated
RUO
Q9Y234
51601
Synthetic peptides corresponding to LIPT1(lipoyltransferase 1) The peptide sequence was selected from the N terminal of LIPT1. Peptide sequence NCFQLLCNCQVPAAGFKKTVKNGLILQSISNDVYQNLAVEDWIHDHMNLE.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.