Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIPT2-AS1 Antibody (OTI4C7), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N271949J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271949J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271949J549 Supplier Novus Biologicals Supplier No. NBP271949JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome c Monoclonal specifically detects Cytochrome c in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cytochrome c
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C7
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbols CYCS
Host Species Mouse
Immunogen Full length recombinant protein of human Cytochrome c (NP_940921) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54205.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.