Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria Monocytogenes Antibody (LZF7), PerCP, Novus Biologicals™
SDP

Catalog No. NB6001281PC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001281PC 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001281PC Supplier Novus Biologicals Supplier No. NB6001281PCP

Mouse Monoclonal Antibody

Listeria Monocytogenes Monoclonal specifically detects Listeria Monocytogenes in Bacteria samples. It is validated for Flow Cytometry.

Specifications

Antigen Listeria Monocytogenes
Applications Flow Cytometry
Classification Monoclonal
Clone LZF7
Conjugate PerCP
Dilution Flow Cytometry 1:10-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias L. monocytogenes
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified L. monocytogenes outer membrane fraction (OM).
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity MAbs recognize Listeria monocytogenes outer membrane fraction and intact L. monocytogenes cells.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.