Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria Monocytogenes Antibody, Novus Biologicals™
SDP

Catalog No. NB6001088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001088 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001088 Supplier Novus Biologicals Supplier No. NB6001088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Listeria Monocytogenes Polyclonal specifically detects Listeria Monocytogenes in Bacteria samples. It is validated for ELISA, Immunofluorescence.

Specifications

Antigen Listeria Monocytogenes
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunofluorescence 1:10-1:500
Formulation 10mM PBS (pH 7.2) with 0.1% Sodium Azide
Gene Alias L. monocytogenes
Host Species Rabbit
Immunogen Listeria monocytogenes; ATCC #43251
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes whole cells of Listeria monocytogenes.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.