Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes antigen 2 Antibody (2:13 C5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309124 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309124 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309124 Supplier Novus Biologicals Supplier No. NBP254650MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Listeria monocytogenes antigen 2 Monoclonal antibody specifically detects Listeria monocytogenes antigen 2 in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Listeria monocytogenes antigen 2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2:13 C5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Listeria monocytogenes antigen 2 protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.