Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes p60 Antibody (p6007), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB173540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173540 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173540 Supplier Novus Biologicals Supplier No. NBP280120AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Listeria monocytogenes p60 Monoclonal antibody specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Listeria monocytogenes p60
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone p6007
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein
Host Species Mouse
Immunogen Recombinant Listeria monocytogenes p60.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.