Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes p60, Mouse anti-Bacteria, Clone: p6007, Novus Biologicals™
SDP

Catalog No. NB013246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013246 50 μg
NB013245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB013246 Supplier Novus Biologicals Supplier No. NBP28012050UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Listeria monocytogenes p60 Monoclonal specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Listeria monocytogenes p60
Applications Western Blot, ELISA
Classification Monoclonal
Clone p6007
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation 0.2um-filtered solution in PBS, pH 7.4.
Gene Alias IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein
Host Species Mouse
Immunogen Recombinant Listeria monocytogenes p60.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Listeria monocytogenes p60 protein. Does not cross-react with other Listeria species.
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.