Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Listeria Selective Oxford Agar Base (Dehydrated)

Catalog No. OXCM0856B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0856B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0856B Supplier Thermo Scientific™ Supplier No. CM0856B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and detect Listeria monocytogenes from pathological, food and environmental samples with this diagnostic medium.

Isolate and detect Listeria monocytogenes from, food and environmental samples with the diagnostic Thermo Scientific™ Oxoid™ Listeria Selective Oxford Agar Base (Dehydrated) when used with Modified Listeria Selective Supplement (Oxford) SR0206. The medium is based on the formulations described by Curtis et al.1 This medium is the specified plating medium in the FDA/BAM isolation procedure2 and in standardized testing methods of other national and international bodies3.

Foodborne infection by Listeria monocytogenes has prompted increased concern for detecting this organism in foods, in the environment and in pathological specimens from both human and animal subjects. The ability to isolate the organism has been impeded in the past by lack of an effective selective medium, as Listeria monocytogenes can be easily and completely overgrown by competing flora.

Use Listeria Selective Oxford Agar Base for selective isolation and detection of Listeria monocytogenes from food and environmental samples.

  • Easy-to-read: Listeria monocytogenes form brown colored colonies with black zones due to aesculin hydrolysis.
  • An FDA/BAM specified plating medium for Listeria monocytogenes isolation3.

The Listeria Selective Oxford Agar Base utilizes the selective inhibitory components and the indicator system―aesculin and ferrous iron for the isolation or differentiation of Listeria monocytogenes. Listeria monocytogenes hydrolyzes the aesculin, producing black zones around the colonies due to the formation of black iron phenolic compounds derived from the aglucon. Gram-negative bacteria are completely inhibited. Most unwanted Gram-positive species are suppressed, but some strains of enterococci grow poorly and exhibit a weak aesculin reaction, usually after 40 hours incubation. Some staphylococci may grow as aesculin-negative colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, IDF, ISO, USDA.
Description Listeria Selective Agar Base (Oxford)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 8.0L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.