Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium carbonate, 99%

Catalog No. AA13418A9
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
10 kg

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1310°C(decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 10 kg
Assay Percent Range 99%
Solubility Information Soluble in water and dilute acids. Insoluble in alcohol.
IUPAC Name dilithium;carbonate
Formula Weight 73.9
Odor Odorless
Appearance White powder
Density 2.11
Chemical Name or Material Lithium carbonate
Show More Show Less
missing translation for 'healthHazard1' H302-H319
missing translation for 'healthHazard3' P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 209-062-5
RTECSNumber OJ5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.