Learn More
Lithium carbonate, 99%
Description
Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1310°C(decomposition) |
| Melting Point | 723°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 99% |
| Solubility Information | Soluble in water and dilute acids. Insoluble in alcohol. |
| IUPAC Name | dilithium(1+) carbonate |
| Formula Weight | 73.9 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302-H319 |
| Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
| EINECSNumber | 209-062-5 |
| RTECSNumber | OJ5800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.