Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium carbonate, 99%, pure

Catalog No. p-222287
Change view
Click to view available options
Quantity:
500 g
10 kg
Packaging:
Plastic bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197785000 500 g Plastic bottle
AC197780100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197785000 Supplier Thermo Scientific Chemicals Supplier No. 197785000
Only null left
Add to Cart

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 720.0°C
Color White
Physical Form Fine Granulated Powder
Quantity 500 g
Assay Percent Range 99%
Linear Formula Li2CO3
Fieser 01,606; 03,183; 04,192; 05,396
Merck Index 15,5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
IUPAC Name dilithium;carbonate
Formula Weight 73.88
Percent Purity 99%
Grade Pure
Loss on Drying 0.4% max. (200°C)
Packaging Plastic bottle
Density 2.11
Chemical Name or Material Lithium carbonate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 209-062-5
RTECSNumber OJ5800000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.