Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hexafluorosilicate, Thermo Scientific Chemicals

Catalog No. p-7050718
Click to view available options
Quantity:
25 g

Lithium hexafluorosilicate is used as chemicals, pharmaceutical intermediates and reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17347-95-4
Molecular Formula F6LiSi
Molecular Weight (g/mol) 149.02
MDL Number MFCD00049473
InChI Key ZXUANORTLYQJEG-UHFFFAOYSA-N

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN2856
Solubility Information Soluble in water and methanol.
Formula Weight 155.96
Odor Odorless
Chemical Name or Material Lithium hexafluorosilicate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROSILICATES, N.O.S.
EINECSNumber 241-372-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.