Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™
Click to view available options
Quantity:
10 g
100 g
Description
Reduction reagent

Specifications
Specifications
Melting Point | 680.0°C |
Color | White to Gray |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 96% min |
Linear Formula | LiH |
Fieser | 04,304; 13,165 |
Merck Index | 15, 5589 |
Solubility Information | Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether |
Formula Weight | 7.95 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. |
Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Handle under inert gas. Protec |
EINECSNumber | 231-484-3 |
RTECSNumber | OJ6300000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction