Alfa Aesar™ Lithium hydroxide monohydrate, 98%

Manufacturer: Alfa Aesar™ A1551922

Quantity 100g

 View more versions of this product

Catalog No. AAA1551922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-66-3
    Molecular Formula LiOH·H2O
    Formula Weight 41.96 (23.95 Anhydrous)
    MDL Number MFCD00149772
    UN Number UN2680
    Merck Index 14,5534
    Chemical Name or Material Lithium hydroxide monohydrate

    Specifications

    Formula Weight 41.96 (23.95 Anhydrous)
    Quantity 100g
    TSCA TSCA
    Molecular Formula LiOH·H2O
    MDL Number MFCD00149772
    CAS 1310-66-3
    UN Number UN2680
    Merck Index 14,5534
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331-H314-H318
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE
    RTECS Number OJ6307080
    Chemical Name or Material Lithium hydroxide monohydrate
    Assay 98%

    Description

    Applications
    Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

    Solubility
    Soluble in water, methanol and ethanol.

    Notes
    Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture, strong oxidizing agents, strong acids and carbon dioxide.

    GHS H Statement
    H301-H331-H314-H318
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit